The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000560.3 | NP_000551.1 | 4 - 5 | 499 | 66 |
NM_001040033.1 | NP_001035122.1 | 5 - 6 | 504 | 66 | |
GenBank mRNA | AK313582.1 | - | 4 - 5 | 452 | 66 |
BC035456.1 | - | 5 - 6 | 486 | 66 | |
BC040693.1 | - | 4 - 5 | 429 | 66 | |
BG536920.1 | - | 4 - 5 | 500 | 66 | |
BT019643.1 | - | 3 - 4 | 333 | 66 | |
CA453414.1 | - | - | 506 | 66 | |
M37033.1 | - | 4 - 5 | 426 | 66 | |
M60871.1 | - | 4 - 5 | 406 | 66 |