Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRXN2
Entrez Gene ID
9379
Gene Name
neurexin 2
Gene Aliases
-
Chromosome Location
Chr.11: 64606174 - 64723188 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015080.3 NP_055895.1 3 - 4 1209 73
XM_005274400.3 XP_005274457.1 2 - 3 762 73
XM_005274401.3 XP_005274458.1 2 - 3 762 73
XM_005274402.3 XP_005274459.1 2 - 3 762 73
XM_011545370.1 XP_011543672.1 2 - 3 762 73
XM_011545373.1 XP_011543675.1 2 - 3 762 73
XM_011545375.1 XP_011543677.1 2 - 3 762 73
XM_017018565.1 XP_016874054.1 2 - 3 762 73
XM_017018567.1 XP_016874056.1 2 - 3 762 73
XM_017018568.1 XP_016874057.1 2 - 3 762 73
XM_017018570.1 XP_016874059.1 2 - 3 762 73
XM_017018571.1 XP_016874060.1 2 - 3 762 73
XM_017018573.1 XP_016874062.1 2 - 3 762 73
XM_017018578.1 XP_016874067.1 2 - 3 762 73
GenBank mRNA AB035266.1 - 2 - 3 861 73

View Full Product Details