Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRXN2
Entrez Gene ID
9379
Gene Name
neurexin 2
Gene Aliases
-
Chromosome Location
Chr.11: 64606174 - 64723188 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138732.2 NP_620060.1 6 - 7 1566 146
XM_005274400.3 XP_005274457.1 6 - 7 1191 146
XM_005274401.3 XP_005274458.1 6 - 7 1191 146
XM_011545373.1 XP_011543675.1 6 - 7 1191 146
GenBank mRNA AB023138.2 - 7 - 8 1543 146
AB177862.1 - 3 - 4 363 146
BC150275.1 - 7 - 8 1543 146

View Full Product Details