The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006204.3 | NP_006195.3 | 2 - 3 | 770 | 114 |
GenBank mRNA | U31973.1 | - | 2 - 3 | 770 | 114 |