The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021620.3 | NP_067633.2 | 2 - 3 | 532 | 71 |
GenBank mRNA | AY004253.1 | - | 2 - 3 | 562 | 71 |