The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286102.1 | NP_001273031.1 | 8 - 9 | 1108 | 62 |
NM_003269.4 | NP_003260.1 | 8 - 9 | 1701 | 62 | |
GenBank mRNA | AB307709.1 | - | 8 - 9 | 993 | 62 |
AF220532.1 | - | 8 - 9 | 1700 | 62 | |
AF411525.1 | - | 8 - 9 | 1234 | 62 | |
AK131541.1 | - | 8 - 9 | 1108 | 62 | |
BC028031.1 | - | 8 - 9 | 1198 | 62 | |
HQ692846.1 | - | 8 - 9 | 993 | 62 | |
Y13276.1 | - | 8 - 9 | 1161 | 62 |