The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318052.1 | NP_001304981.1 | 17 - 18 | 3328 | 93 |
NM_015689.4 | NP_056504.3 | 18 - 19 | 3430 | 93 | |
NR_134477.1 | 16 - 17 | 2997 | 93 | ||
XM_005249977.3 | XP_005250034.1 | 18 - 19 | 3961 | 93 | |
XM_011516052.2 | XP_011514354.1 | 18 - 19 | 3308 | 93 | |
XM_011516053.2 | XP_011514355.1 | 17 - 18 | 3412 | 93 | |
XM_011516054.2 | XP_011514356.1 | 18 - 19 | 3213 | 93 | |
XM_017011989.1 | XP_016867478.1 | 17 - 18 | 3414 | 93 | |
GenBank mRNA | AB033103.2 | - | 17 - 18 | 3328 | 93 |
AK290056.1 | - | 18 - 19 | 3199 | 93 | |
AK297959.1 | - | 10 - 11 | 2089 | 93 | |
BC049193.1 | - | 14 - 15 | 1798 | 93 | |
BC132875.1 | - | 16 - 17 | 3025 | 93 |