The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172167.2 | NP_751907.1 | 2 - 3 | 514 | 96 |
NM_172168.2 | NP_751908.1 | 2 - 3 | 514 | 96 | |
XM_005255099.4 | XP_005255156.1 | 3 - 4 | 1609 | 96 | |
XM_017022927.1 | XP_016878416.1 | 3 - 4 | 1115 | 96 | |
XM_017022928.1 | XP_016878417.1 | 3 - 4 | 472 | 96 | |
GenBank mRNA | AB097667.1 | - | 2 - 3 | 324 | 96 |
AF532984.1 | - | 2 - 3 | 148 | 96 | |
AF532985.1 | - | 2 - 3 | 148 | 96 | |
AF539796.1 | - | 2 - 3 | 144 | 96 | |
AK292197.1 | - | 2 - 3 | 514 | 96 | |
AK313917.1 | - | 2 - 3 | 434 | 96 |