The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033033.3 | NP_149022.3 | 4 - 5 | 895 | 74 |
GenBank mRNA | AJ628419.1 | - | 4 - 5 | 858 | 74 |