Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SOHLH1
Entrez Gene ID
402381
Gene Name
spermatogenesis and oogenesis specific basic helix-loop-helix 1
Gene Aliases
C9orf157, NOHLH, SPATA27, TEB2, bA100C15.3, bHLHe80
Chromosome Location
Chr.9: 135691862 - 135701316 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012415.2 NP_001012415.2 6 - 7 936 66
NM_001101677.1 NP_001095147.1 6 - 7 936 66
XM_005263403.3 XP_005263460.1 6 - 7 887 66
XM_006717109.3 XP_006717172.1 7 - 8 2012 66
XM_011518698.2 XP_011517000.1 8 - 9 2041 66
GenBank mRNA AY902244.1 - 6 - 7 936 66
BC031861.1 - 6 - 7 947 66
EB710265.1 - 4 - 5 633 66

View Full Product Details