Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB41
Entrez Gene ID
360023
Gene Name
zinc finger and BTB domain containing 41
Gene Aliases
FRBZ1, ZNF924
Chromosome Location
Chr.1: 197153682 - 197200542 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_194314.2 NP_919290.2 7 - 8 1942 89
XM_011509507.2 XP_011507809.1 7 - 8 1921 89
GenBank mRNA AY163816.2 - 7 - 8 1942 89
AY336933.1 - 4 - 5 413 89
BC132818.1 - 7 - 8 1906 89
BC136600.1 - 7 - 8 1906 89

View Full Product Details