The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_037584.2 | 2 - 3 | 222 | 68 | |
GenBank mRNA | AF381171.1 | - | 2 - 3 | 188 | 68 |
AF381172.1 | - | 2 - 3 | 191 | 68 | |
AF381173.1 | - | 2 - 3 | 198 | 68 | |
AF381174.1 | - | 2 - 3 | 198 | 68 |