Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MOXD2P
- Entrez Gene ID
-
100289017
- Gene Name
- monooxygenase, DBH-like 2, pseudogene
- Gene Aliases
-
MOXD2
- Chromosome Location
-
Chr.7: 142240737 - 142247067 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_024346.3 |
|
1 - 2 |
292 |
80 |
View Full Product Details