The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130136.1 | NP_001123608.1 | 1 - 2 | 587 | 63 |
GenBank mRNA | AB040898.2 | - | 1 - 2 | 587 | 63 |
AK092302.1 | - | 1 - 2 | 603 | 63 | |
BC152429.1 | - | 1 - 2 | 587 | 63 |