Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHIC1
Entrez Gene ID
53344
Gene Name
cysteine rich hydrophobic domain 1
Gene Aliases
BRX
Chromosome Location
Chr.X: 73563045 - 73687109 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039840.2 NP_001034929.2 3 - 4 644 103
XM_017029583.1 XP_016885072.1 4 - 5 325 103
GenBank mRNA AK297768.1 - 3 - 4 501 103
AL832095.1 - 3 - 4 284 103
BC127728.1 - 3 - 4 555 103
BC136691.1 - 3 - 4 549 103
BC136692.1 - 3 - 4 549 103

View Full Product Details