Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MTX3
Entrez Gene ID
345778
Gene Name
metaxin 3
Gene Aliases
-
Chromosome Location
Chr.5: 79976716 - 79991267 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001010891.4 NP_001010891.4 4 - 5 345 138
NM_001167741.1 NP_001161213.1 3 - 4 275 138
XM_005248495.3 XP_005248552.1 4 - 5 347 138
XM_017009440.1 XP_016864929.1 3 - 4 269 138
GenBank mRNA AK296864.1 - 4 - 5 340 138
BC150295.1 - 4 - 5 335 138
BX538064.1 - 3 - 4 255 138
BX647596.1 - 4 - 5 342 138
DB456588.1 - 4 - 5 345 138

View Full Product Details