Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRDM6
Entrez Gene ID
93166
Gene Name
PR domain 6
Gene Aliases
PRISM
Chromosome Location
Chr.5: 123089146 - 123194266 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136239.1 NP_001129711.1 4 - 5 1409 84
XM_011543726.2 XP_011542028.1 3 - 4 1472 84
GenBank mRNA AF272898.1 - 4 - 5 1059 84
AK297992.1 - 3 - 4 1025 84

View Full Product Details