Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPEG
Entrez Gene ID
10290
Gene Name
SPEG complex locus
Gene Aliases
APEG-1, APEG1, BPEG, CNM5, SPEGalpha, SPEGbeta
Chromosome Location
Chr.2: 219434215 - 219493632 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005876.4 NP_005867.3 34 - 35 8143 63
XM_005246237.2 XP_005246294.1 36 - 37 7871 63
XM_005246239.2 XP_005246296.1 36 - 37 7686 63
XM_005246240.2 XP_005246297.1 29 - 30 5888 63
XM_005246241.1 XP_005246298.1 29 - 30 5782 63
XM_005246242.4 XP_005246299.1 29 - 30 5907 63
XM_006712189.3 XP_006712252.1 35 - 36 7876 63
XM_006712193.3 XP_006712256.1 29 - 30 9007 63
XM_011510479.2 XP_011508781.1 35 - 36 8309 63
XM_011510483.2 XP_011508785.2 35 - 36 7906 63
XM_017003157.1 XP_016858646.1 35 - 36 7906 63
XM_017003158.1 XP_016858647.1 31 - 32 7482 63
XM_017003160.1 XP_016858649.1 19 - 20 4078 63
GenBank mRNA AB037718.1 - 25 - 26 5200 63
AY603755.1 - 33 - 34 7756 63

View Full Product Details