Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPEG
Entrez Gene ID
10290
Gene Name
SPEG complex locus
Gene Aliases
APEG-1, APEG1, BPEG, CNM5, SPEGalpha, SPEGbeta
Chromosome Location
Chr.2: 219434215 - 219493632 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005876.4 NP_005867.3 11 - 12 3305 61
XM_005246237.2 XP_005246294.1 13 - 14 3033 61
XM_005246239.2 XP_005246296.1 13 - 14 2848 61
XM_005246240.2 XP_005246297.1 6 - 7 1050 61
XM_005246241.1 XP_005246298.1 6 - 7 944 61
XM_005246242.4 XP_005246299.1 6 - 7 1069 61
XM_006712189.3 XP_006712252.1 12 - 13 3038 61
XM_006712193.3 XP_006712256.1 6 - 7 4169 61
XM_011510479.2 XP_011508781.1 12 - 13 3471 61
XM_011510483.2 XP_011508785.2 12 - 13 3068 61
XM_017003157.1 XP_016858646.1 12 - 13 3068 61
XM_017003158.1 XP_016858647.1 8 - 9 2644 61
XM_017003159.1 XP_016858648.1 9 - 10 2827 61
GenBank mRNA AB037718.1 - 2 - 3 362 61
AY603755.1 - 10 - 11 2918 61

View Full Product Details