The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005876.4 | NP_005867.3 | 16 - 17 | 4144 | 126 |
XM_005246237.2 | XP_005246294.1 | 18 - 19 | 3872 | 126 | |
XM_005246239.2 | XP_005246296.1 | 18 - 19 | 3687 | 126 | |
XM_005246240.2 | XP_005246297.1 | 11 - 12 | 1889 | 126 | |
XM_005246241.1 | XP_005246298.1 | 11 - 12 | 1783 | 126 | |
XM_005246242.4 | XP_005246299.1 | 11 - 12 | 1908 | 126 | |
XM_006712189.3 | XP_006712252.1 | 17 - 18 | 3877 | 126 | |
XM_006712193.3 | XP_006712256.1 | 11 - 12 | 5008 | 126 | |
XM_011510479.2 | XP_011508781.1 | 17 - 18 | 4310 | 126 | |
XM_011510483.2 | XP_011508785.2 | 17 - 18 | 3907 | 126 | |
XM_017003157.1 | XP_016858646.1 | 17 - 18 | 3907 | 126 | |
XM_017003158.1 | XP_016858647.1 | 13 - 14 | 3483 | 126 | |
XM_017003159.1 | XP_016858648.1 | 14 - 15 | 3666 | 126 | |
XM_017003160.1 | XP_016858649.1 | 1 - 2 | 90 | 126 | |
GenBank mRNA | AB037718.1 | - | 7 - 8 | 1201 | 126 |
AY603755.1 | - | 15 - 16 | 3757 | 126 |