The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198137.1 | NP_937770.1 | 4 - 5 | 557 | 95 |
XM_011541432.2 | XP_011539734.1 | 4 - 5 | 664 | 95 | |
XM_011541433.2 | XP_011539735.1 | 4 - 5 | 1223 | 95 | |
GenBank mRNA | BC128138.1 | - | 4 - 5 | 556 | 95 |
BC128139.1 | - | 4 - 5 | 557 | 95 |