The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011511673.2 | XP_011509975.1 | 8 - 9 | 1430 | 148 |
XM_011511677.2 | XP_011509979.1 | 7 - 8 | 1330 | 148 | |
XM_011511678.2 | XP_011509980.1 | 3 - 4 | 928 | 148 | |
GenBank mRNA | BC041460.1 | - | 4 - 5 | 2685 | 148 |