The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010978.3 | NP_001010978.2 | 1 - 2 | 98 | 68 |
NM_001276393.1 | NP_001263322.1 | 1 - 2 | 98 | 68 | |
NM_001276394.1 | NP_001263323.1 | 1 - 2 | 98 | 68 | |
NM_001276395.1 | NP_001263324.1 | 1 - 2 | 98 | 68 | |
XM_011541446.1 | XP_011539748.1 | 1 - 2 | 81 | 68 | |
XM_017001264.1 | XP_016856753.1 | 1 - 2 | 81 | 68 | |
GenBank mRNA | BC103740.1 | - | 1 - 2 | 86 | 68 |
BC127708.1 | - | 1 - 2 | 95 | 68 | |
BC140760.1 | - | 1 - 2 | 42 | 68 | |
BC144466.1 | - | 1 - 2 | 42 | 68 | |
BC171743.1 | - | 1 - 2 | 42 | 68 | |
CF887342.1 | - | 1 - 2 | 98 | 68 |