The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145444.1 | NP_001138916.1 | 16 - 17 | 2594 | 86 |
NM_001145445.1 | NP_001138917.1 | 17 - 18 | 2639 | 86 | |
NM_020706.2 | NP_065757.1 | 17 - 18 | 2639 | 86 | |
XM_005261017.2 | XP_005261074.1 | 16 - 17 | 2443 | 86 | |
XM_006724035.2 | XP_006724098.1 | 17 - 18 | 2608 | 86 | |
XM_006724036.2 | XP_006724099.1 | 17 - 18 | 2608 | 86 | |
XM_017028415.1 | XP_016883904.1 | 17 - 18 | 2597 | 86 | |
XM_017028416.1 | XP_016883905.1 | 17 - 18 | 2482 | 86 | |
XM_017028417.1 | XP_016883906.1 | 16 - 17 | 2443 | 86 | |
GenBank mRNA | AB032998.1 | - | 11 - 12 | 1627 | 86 |
AF023142.1 | - | 17 - 18 | 2243 | 86 | |
AK025748.1 | - | 12 - 13 | 1658 | 86 | |
AK027017.1 | - | 3 - 4 | 450 | 86 | |
AK308406.1 | - | 16 - 17 | 2445 | 86 | |
AL117417.1 | - | 12 - 13 | 1647 | 86 | |
AL834304.1 | - | 17 - 18 | 2419 | 86 | |
BC014921.1 | - | 17 - 18 | 2616 | 86 | |
BC043353.1 | - | 17 - 18 | 2596 | 86 | |
BC052286.1 | - | 17 - 18 | 2470 | 86 | |
BC064990.1 | - | 17 - 18 | 2470 | 86 |