The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145444.1 | NP_001138916.1 | 17 - 18 | 2684 | 102 |
NM_020706.2 | NP_065757.1 | 18 - 19 | 2729 | 102 | |
XM_005261017.2 | XP_005261074.1 | 17 - 18 | 2533 | 102 | |
XM_006724035.2 | XP_006724098.1 | 18 - 19 | 2698 | 102 | |
XM_017028415.1 | XP_016883904.1 | 18 - 19 | 2687 | 102 | |
GenBank mRNA | AF023142.1 | - | 18 - 19 | 2333 | 102 |
AK308406.1 | - | 17 - 18 | 2535 | 102 | |
AL117417.1 | - | 13 - 14 | 1737 | 102 | |
BC014921.1 | - | 18 - 19 | 2706 | 102 | |
BC043353.1 | - | 18 - 19 | 2686 | 102 | |
BC052286.1 | - | 18 - 19 | 2560 | 102 | |
BC064990.1 | - | 18 - 19 | 2560 | 102 |