The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145444.1 | NP_001138916.1 | 18 - 19 | 2871 | 77 |
NM_001145445.1 | NP_001138917.1 | 19 - 20 | 2850 | 77 | |
NM_020706.2 | NP_065757.1 | 19 - 20 | 2916 | 77 | |
XM_005261017.2 | XP_005261074.1 | 18 - 19 | 2720 | 77 | |
XM_006724035.2 | XP_006724098.1 | 19 - 20 | 2885 | 77 | |
XM_006724036.2 | XP_006724099.1 | 19 - 20 | 2819 | 77 | |
XM_017028415.1 | XP_016883904.1 | 19 - 20 | 2874 | 77 | |
XM_017028416.1 | XP_016883905.1 | 19 - 20 | 2693 | 77 | |
XM_017028417.1 | XP_016883906.1 | 18 - 19 | 2654 | 77 | |
GenBank mRNA | AB032998.1 | - | 13 - 14 | 1838 | 77 |
AK025748.1 | - | 14 - 15 | 1869 | 77 | |
AK027017.1 | - | 5 - 6 | 661 | 77 | |
AK308406.1 | - | 18 - 19 | 2722 | 77 | |
AL834304.1 | - | 19 - 20 | 2630 | 77 | |
BC014921.1 | - | 19 - 20 | 2893 | 77 | |
BC043353.1 | - | 19 - 20 | 2873 | 77 | |
BC052286.1 | - | 19 - 20 | 2747 | 77 | |
BC064990.1 | - | 19 - 20 | 2747 | 77 |