Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCAF4
Entrez Gene ID
57466
Gene Name
SR-related CTD associated factor 4
Gene Aliases
SFRS15, SRA4
Chromosome Location
Chr.21: 31671000 - 31732118 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145444.1 NP_001138916.1 5 - 6 982 64
NM_001145445.1 NP_001138917.1 6 - 7 1027 64
NM_020706.2 NP_065757.1 6 - 7 1027 64
XM_006724035.2 XP_006724098.1 6 - 7 1008 64
XM_006724036.2 XP_006724099.1 6 - 7 1008 64
XM_017028415.1 XP_016883904.1 6 - 7 988 64
XM_017028416.1 XP_016883905.1 6 - 7 873 64
GenBank mRNA AF023142.1 - 6 - 7 631 64
AK025748.1 - 1 - 2 46 64
AK057840.1 - 6 - 7 870 64
AK308406.1 - 5 - 6 833 64
AL117417.1 - 1 - 2 49 64
AL834304.1 - 6 - 7 807 64
BC014921.1 - 6 - 7 1004 64
BC043353.1 - 6 - 7 984 64
BC052286.1 - 6 - 7 858 64
BC064990.1 - 6 - 7 858 64

View Full Product Details