The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198147.2 | NP_937790.2 | 1 - 2 | 938 | 94 |
GenBank mRNA | AK056717.1 | - | 1 - 2 | 451 | 94 |
AY358212.1 | - | 1 - 2 | 944 | 94 | |
BC012476.1 | - | 1 - 2 | 938 | 94 |