Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PEAR1
Entrez Gene ID
375033
Gene Name
platelet endothelial aggregation receptor 1
Gene Aliases
JEDI, MEGF12
Chromosome Location
Chr.1: 156893697 - 156916434 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080471.1 NP_001073940.1 22 - 23 3084 120
XM_005245141.3 XP_005245198.1 23 - 24 3277 120
XM_011509510.2 XP_011507812.1 23 - 24 3216 120
XM_011509511.2 XP_011507813.1 22 - 23 3172 120
XM_011509512.2 XP_011507814.1 22 - 23 3096 120
XM_017001234.1 XP_016856723.1 22 - 23 4342 120
XM_017001235.1 XP_016856724.1 22 - 23 4212 120
XM_017001236.1 XP_016856725.1 22 - 23 3179 120
XM_017001237.1 XP_016856726.1 23 - 24 3423 120
XM_017001238.1 XP_016856727.1 23 - 24 3362 120
XM_017001239.1 XP_016856728.1 23 - 24 3173 120
XM_017001240.1 XP_016856729.1 20 - 21 2741 120
XM_017001241.1 XP_016856730.1 20 - 21 2889 120
XM_017001242.1 XP_016856731.1 20 - 21 2793 120
GenBank mRNA AK074121.1 - 17 - 18 2696 120

View Full Product Details