The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080471.1 | NP_001073940.1 | 14 - 15 | 1936 | 74 |
XM_005245141.3 | XP_005245198.1 | 15 - 16 | 2129 | 74 | |
XM_011509510.2 | XP_011507812.1 | 15 - 16 | 2068 | 74 | |
XM_011509511.2 | XP_011507813.1 | 14 - 15 | 2024 | 74 | |
XM_011509512.2 | XP_011507814.1 | 14 - 15 | 1948 | 74 | |
XM_017001234.1 | XP_016856723.1 | 14 - 15 | 3194 | 74 | |
XM_017001235.1 | XP_016856724.1 | 14 - 15 | 3064 | 74 | |
XM_017001236.1 | XP_016856725.1 | 14 - 15 | 2031 | 74 | |
XM_017001237.1 | XP_016856726.1 | 15 - 16 | 2275 | 74 | |
XM_017001238.1 | XP_016856727.1 | 15 - 16 | 2214 | 74 | |
XM_017001239.1 | XP_016856728.1 | 15 - 16 | 2025 | 74 | |
XM_017001240.1 | XP_016856729.1 | 12 - 13 | 1593 | 74 | |
XM_017001241.1 | XP_016856730.1 | 12 - 13 | 1741 | 74 | |
XM_017001242.1 | XP_016856731.1 | 12 - 13 | 1645 | 74 | |
GenBank mRNA | AK074121.1 | - | 9 - 10 | 1548 | 74 |