Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TWISTNB
Entrez Gene ID
221830
Gene Name
TWIST neighbor
Gene Aliases
-
Chromosome Location
Chr.7: 19695462 - 19709037 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002926.1 NP_001002926.1 2 - 3 415 96
GenBank mRNA AK301329.1 - 2 - 3 439 96
BC014574.1 - 2 - 3 417 96
BC056267.1 - 2 - 3 429 96
BC130298.1 - 2 - 3 414 96
BC130300.1 - 2 - 3 413 96

View Full Product Details