The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_194285.2 | NP_919261.2 | 1 - 2 | 274 | 63 |
GenBank mRNA | AK096760.1 | - | 1 - 2 | 294 | 63 |
AK304078.1 | - | 1 - 2 | 190 | 63 | |
BC056261.1 | - | 1 - 2 | 152 | 63 | |
BX641102.1 | - | 1 - 2 | 174 | 63 | |
BX647798.1 | - | 1 - 2 | 274 | 63 | |
BX648478.1 | - | 1 - 2 | 196 | 63 |