The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142649.1 | NP_001136121.1 | 14 - 15 | 1719 | 79 |
NM_213599.2 | NP_998764.1 | 14 - 15 | 1722 | 79 | |
XM_005252820.3 | XP_005252877.2 | 13 - 14 | 1826 | 79 | |
XM_005252821.3 | XP_005252878.2 | 13 - 14 | 1823 | 79 | |
XM_005252822.4 | XP_005252879.1 | 14 - 15 | 1607 | 79 | |
XM_011519949.2 | XP_011518251.1 | 12 - 13 | 1775 | 79 | |
GenBank mRNA | AB125267.1 | - | 14 - 15 | 1722 | 79 |
AL832374.1 | - | 14 - 15 | 1744 | 79 | |
AL833271.1 | - | 14 - 15 | 1743 | 79 |