Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- NAV2-AS4
- Entrez Gene ID
-
399876
- Gene Name
- NAV2 antisense RNA 4
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 19510893 - 19524078 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_033989.1 |
|
2 - 3 |
333 |
69 |
GenBank mRNA |
AK023501.1 |
- |
2 - 3 |
333 |
69 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AU135729.1 |
- |
2 - 3 |
333 |
|
Chr. 11 - 19511403 - 19524078 [-] on Build GRCh38 |
69 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
NAV2 antisense RNA 5 |
NAV2-AS5 |
|
-11.2kb
|
Sense |
Chr. - 19502672 - 19507229 on Build GRCh38 |
Downstream |
microRNA 4486 |
MIR4486 |
|
55.4kb
|
Anti-sense |
Chr. - 19575310 - 19575372 on Build GRCh38 |
View Full Product Details