Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LOC100507654
- Entrez Gene ID
-
100507654
- Gene Name
- uncharacterized LOC100507654
- Gene Aliases
-
-
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC034289.1 |
- |
3 - 4 |
226 |
76 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BG721018.1 |
- |
3 - 4 |
231 |
|
Chr. 13 - 100537323 - 100580113 [-] on Build GRCh38 |
76 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
gamma-glutamylamine cyclotransferase |
GGACT |
A2LD1 |
|
Sense |
Chr. - 100530164 - 100588792 on Build GRCh38 |
Downstream |
transmembrane and tetratricopeptide repeat containing 4 |
TMTC4 |
|
62.0kb
|
Sense |
Chr. - 100603836 - 100675025 on Build GRCh38 |
View Full Product Details