The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080826.1 | NP_543016.1 | 5 - 6 | 885 | 70 |
GenBank mRNA | AK307409.1 | - | 3 - 4 | 324 | 70 |
DB202450.1 | - | 3 - 4 | 324 | 70 |