Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZFP62
Entrez Gene ID
643836
Gene Name
ZFP62 zinc finger protein
Gene Aliases
ZET, ZNF755
Chromosome Location
Chr.5: 180843580 - 180861327 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172638.1 NP_001166109.1 1 - 2 73 66
XM_017009720.1 XP_016865209.1 1 - 2 114 66
GenBank mRNA AK303450.1 - 1 - 2 64 66

View Full Product Details