The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278562.1 | NP_001265491.1 | 1 - 2 | 129 | 68 |
GenBank mRNA | AK090598.1 | - | 1 - 2 | 114 | 68 |