Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LCTL
Entrez Gene ID
197021
Gene Name
lactase like
Gene Aliases
KLG, KLPH
Chromosome Location
Chr.15: 66547468 - 66566025 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278562.1 NP_001265491.1 8 - 9 1033 131
NM_207338.3 NP_997221.2 9 - 10 1324 131
NR_103739.1 7 - 8 1046 131
XM_017021999.1 XP_016877488.1 9 - 10 1243 131
GenBank mRNA AK090598.1 - 8 - 9 1018 131
AK311441.1 - 7 - 8 1046 131
AY358729.1 - 9 - 10 1324 131

View Full Product Details