Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTGFRN
Entrez Gene ID
5738
Gene Name
prostaglandin F2 receptor inhibitor
Gene Aliases
CD315, CD9P-1, EWI-F, FPRP, SMAP-6
Chromosome Location
Chr.1: 116909923 - 116990358 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020440.3 NP_065173.2 4 - 5 1496 135
XM_017001874.1 XP_016857363.1 4 - 5 1267 135
GenBank mRNA AB037857.1 - 4 - 5 1352 135
AK074637.1 - 4 - 5 1362 135
BC098142.1 - 4 - 5 942 135
BC114520.1 - 3 - 4 793 135
BC139718.1 - 3 - 4 539 135
BC152454.1 - 4 - 5 1352 135

View Full Product Details