The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175078.2 | NP_778253.2 | 4 - 5 | 945 | 92 |
XM_011538288.2 | XP_011536590.1 | 4 - 5 | 707 | 92 | |
GenBank mRNA | BC118598.1 | - | 5 - 6 | 1030 | 92 |
BC122558.1 | - | 4 - 5 | 945 | 92 |