Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRK
Entrez Gene ID
203447
Gene Name
Nik related kinase
Gene Aliases
NESK
Chromosome Location
Chr.X: 105821944 - 105958610 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198465.3 NP_940867.2 23 - 24 4260 119
XM_006724632.3 XP_006724695.1 23 - 24 4222 119
XM_006724633.3 XP_006724696.1 23 - 24 4222 119
XM_006724634.3 XP_006724697.1 23 - 24 4222 119
XM_011530887.2 XP_011529189.1 23 - 24 4014 119
GenBank mRNA BX538345.1 - 23 - 24 4262 119

View Full Product Details