The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320870.1 | NP_001307799.1 | 4 - 5 | 1264 | 63 |
NM_001320877.1 | NP_001307806.1 | 3 - 4 | 1198 | 63 | |
NR_135483.1 | 4 - 5 | 1264 | 63 | ||
NR_135484.1 | 4 - 5 | 1264 | 63 | ||
XM_005256641.2 | XP_005256698.1 | 3 - 4 | 1168 | 63 | |
XM_011523846.1 | XP_011522148.1 | 4 - 5 | 1321 | 63 | |
XM_017024633.1 | XP_016880122.1 | 4 - 5 | 2191 | 63 | |
XM_017024634.1 | XP_016880123.1 | 4 - 5 | 2485 | 63 | |
GenBank mRNA | AY498866.1 | - | 4 - 5 | 728 | 63 |
BC101330.2 | - | 4 - 5 | 757 | 63 | |
BC105995.1 | - | 4 - 5 | 1239 | 63 |