The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_125367.1 | 6 - 7 | 556 | 138 | |
GenBank mRNA | AK097500.1 | - | 6 - 7 | 544 | 138 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | myosin heavy chain 2 | MYH2 | IBM3, MYH2A, MYHSA2, MYHas8, MYPOP, MyHC-2A, MyHC-IIa | -48.4kb | Anti-sense | Chr. - 10521148 - 10549700 on Build GRCh38 |
Downstream | myosin, heavy chain 3, skeletal muscle, embryonic | MYH3 | DA2A, DA2B, DA8, HEMHC, MYHC-EMB, MYHSE1, SMHCE | 20.3kb | Anti-sense | Chr. - 10628526 - 10679657 on Build GRCh38 |