Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VASN
Entrez Gene ID
114990
Gene Name
vasorin
Gene Aliases
SLITL2
Chromosome Location
Chr.16: 4371848 - 4383528 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138440.2 NP_612449.2 1 - 2 150 129
GenBank mRNA AY358298.1 - 1 - 2 131 129
AY358299.1 - 1 - 2 114 129
BC068575.1 - 1 - 2 150 129

View Full Product Details