Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACSM4
Entrez Gene ID
341392
Gene Name
acyl-CoA synthetase medium-chain family member 4
Gene Aliases
-
Chromosome Location
Chr.12: 7304332 - 7328373 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080454.1 NP_001073923.1 10 - 11 1436 62

View Full Product Details