The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001247989.1 | NP_001234918.1 | 5 - 6 | 740 | 69 |
NM_001247990.1 | NP_001234919.1 | 5 - 6 | 630 | 69 | |
GenBank mRNA | AK055791.1 | - | 5 - 6 | 860 | 69 |
BC051363.1 | - | 5 - 6 | 343 | 69 | |
BC064355.1 | - | 5 - 6 | 686 | 69 |