The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_040100.1 | 2 - 3 | 262 | 122 | |
GenBank mRNA | BC047546.1 | - | 2 - 3 | 259 | 122 |
BG527749.1 | - | 2 - 3 | 135 | 122 |