The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020759.2 | NP_065810.2 | 28 - 29 | 13554 | 84 |
XM_011521831.2 | XP_011520133.1 | 28 - 29 | 13567 | 84 | |
XM_011521832.2 | XP_011520134.1 | 28 - 29 | 13564 | 84 | |
XM_011521833.2 | XP_011520135.1 | 27 - 28 | 13534 | 84 | |
XM_011521834.2 | XP_011520136.1 | 27 - 28 | 13504 | 84 | |
XM_011521835.2 | XP_011520137.1 | 26 - 27 | 13426 | 84 | |
XM_011521836.2 | XP_011520138.1 | 28 - 29 | 13869 | 84 | |
XM_011521837.2 | XP_011520139.1 | 26 - 27 | 13336 | 84 | |
XM_011521839.2 | XP_011520141.1 | 15 - 16 | 12336 | 84 | |
XM_017022439.1 | XP_016877928.1 | 27 - 28 | 13489 | 84 | |
XM_017022440.1 | XP_016877929.1 | 27 - 28 | 13466 | 84 | |
XM_017022441.1 | XP_016877930.1 | 26 - 27 | 13785 | 84 | |
XM_017022442.1 | XP_016877931.1 | 21 - 22 | 12893 | 84 | |
XM_017022443.1 | XP_016877932.1 | 20 - 21 | 12877 | 84 | |
GenBank mRNA | AB037721.1 | - | 6 - 7 | 4858 | 84 |
AK297603.1 | - | 6 - 7 | 1074 | 84 | |
AL133579.1 | - | 2 - 3 | 201 | 84 | |
CR749416.1 | - | 6 - 7 | 7576 | 84 | |
CR936665.1 | - | 6 - 7 | 5549 | 84 |