The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100600.1 | NP_001094070.1 | 6 - 7 | 805 | 58 |
NM_198403.3 | NP_940685.3 | 6 - 7 | 733 | 58 | |
NR_072989.1 | 7 - 8 | 859 | 58 | ||
GenBank mRNA | AK094524.1 | - | 7 - 8 | 857 | 58 |
AY424288.1 | - | 6 - 7 | 642 | 58 | |
BC037881.1 | - | 6 - 7 | 756 | 58 | |
BC067905.1 | - | 6 - 7 | 733 | 58 |